siRNA Details
Gene Symbol: Oosp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133353.3 | NP_579931.1 | 5 | 602 | ||
GenBank mRNA | AB050982.2 | - | 5 | 595 | ||
AB086437.1 | - | 4 | 487 | |||
AF420487.1 | - | 5 | 607 | |||
AK033038.1 | - | 5 | 605 | |||
AK162132.1 | - | 5 | 605 | |||
BC100313.1 | - | 5 | 581 | 30936071 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
