siRNA Details
Gene Symbol: Tm4sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008536.3 | NP_032562.1 | 6 | 1149 | ||
GenBank mRNA | AK077326.1 | - | 4 | 652 | ||
AK134603.1 | - | 5 | 1085 | |||
AK164718.1 | - | 4 | 653 | |||
BC083073.1 | - | 6 | 1149 | 6337376 | ||
KC508089.1 | - | 4 | 624 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
