siRNA Details
Gene Symbol: Smad3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016769.4 | NP_058049.3 | 1 | 495 | ||
GenBank mRNA | AB008192.1 | - | 1 | 204 | ||
AF016189.1 | - | 1 | 471 | |||
AK041743.1 | - | 1 | 16 | |||
AK048626.1 | - | 1 | 495 | |||
AK083158.1 | - | 1 | 103 | |||
BC022904.1 | - | 1 | 638 | |||
BC066850.1 | - | 1 | 181 | 30432720 |

More Information
Other Species Targeted

Gene Ontology Categories:
