siRNA Details
Gene Symbol: Ascl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008553.4 | NP_032579.2 | 2 | 2087 | ||
GenBank mRNA | AK143210.1 | - | 2 | 2088 | ||
BC055748.1 | - | 2 | 1608 | 6415061 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
