siRNA Details
Gene Symbol: Mbd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF072248.1 | - | 4 | 710 | ||
BC038264.1 | - | 4 | 511 | 5250237 | ||
BQ042794.1 | - | 3 | 278 | |||
CK625083.1 | - | 4 | 530 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
