siRNA Details
Gene Symbol: Foxc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008592.2 | NP_032618.2 | 1 | 1257 | ||
GenBank mRNA | AF045017.1 | - | 1 | 1244 | ||
AK144420.1 | - | 1 | 1160 | |||
BC052011.1 | - | 1 | 1095 | 5720391 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
