siRNA Details
Gene Symbol: Ndrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008681.2 | NP_032707.2 | 10 | 826 | ||
GenBank mRNA | AA795988.1 | - | 2 | 119 | ||
AK004548.1 | - | 10 | 826 | |||
AK098075.1 | - | 10 | 825 | |||
AK161860.1 | - | 10 | 828 | |||
AK165007.1 | - | 10 | 826 | |||
AK170281.1 | - | 10 | 827 | |||
AK170636.1 | - | 10 | 825 | |||
AK170809.1 | - | 10 | 826 | |||
BC015282.1 | - | 10 | 781 | 4237943 | ||
BC071235.1 | - | 10 | 834 | 4242075 | ||
BC141246.1 | - | 10 | 717 | 9055858 | ||
BC171968.1 | - | 10 | 717 | 9054254 | ||
CA320478.1 | - | 7 | 569 | |||
U52073.1 | - | 10 | 799 | |||
U60593.1 | - | 10 | 824 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
