siRNA Details
Gene Symbol: Ndrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008681.2 | NP_032707.2 | 12, 13 | 971 | ||
GenBank mRNA | AA795988.1 | - | 4, 5 | 264 | ||
AK004548.1 | - | 12, 13 | 971 | |||
AK098075.1 | - | 12, 13 | 970 | |||
AK161860.1 | - | 12, 13 | 973 | |||
AK165007.1 | - | 12, 13 | 971 | |||
AK170281.1 | - | 12, 13 | 972 | |||
AK170439.1 | - | 12, 13 | 970 | |||
AK170636.1 | - | 12, 13 | 970 | |||
AK170809.1 | - | 12, 13 | 971 | |||
BC015282.1 | - | 12, 13 | 926 | 4237943 | ||
BC071235.1 | - | 12, 13 | 979 | 4242075 | ||
BC141246.1 | - | 12, 13 | 862 | 9055858 | ||
U52073.1 | - | 12, 13 | 944 | |||
U60593.1 | - | 12, 13 | 969 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
