siRNA Details
Gene Symbol: Nfix | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081981.2 | NP_001075450.1 | 2 | 1389 | ||
NM_001081982.2 | NP_001075451.1 | 2 | 714 | |||
NM_001297601.1 | NP_001284530.1 | 2 | 1389 | |||
NM_010906.3 | NP_035036.1 | 2 | 714 | |||
GenBank mRNA | AK158568.1 | - | 2 | 1392 | ||
BC003766.1 | - | 2 | 436 | 3491917 | ||
BC090626.1 | - | 2 | 416 | |||
S81451.1 | - | 1 | 400 | |||
U57636.1 | - | 1 | 391 | |||
Y07688.1 | - | 2 | 716 | |||
Y07689.1 | - | 2 | 518 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
