siRNA Details
Gene Symbol: Slc11a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013612.2 | NP_038640.2 | 12 | 1388 | ||
GenBank mRNA | AK090303.1 | - | 11 | 1668 | ||
AK144237.1 | - | 12 | 1373 | |||
AK171343.1 | - | 9 | 1813 | |||
AK171393.1 | - | 12 | 1369 | |||
BC109137.1 | - | 12 | 1302 | 40039390 | ||
BC109138.1 | - | 12 | 1302 | 40039397 | ||
L13732.1 | - | 12 | 1366 | |||
X75355.1 | - | 12 | 1347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
