siRNA Details
Gene Symbol: Nrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190187.1 | NP_001177116.1 | 1, 2 | 1118 | ||
NM_001190188.1 | NP_001177117.1 | 1, 2 | 1118 | |||
NM_008734.3 | NP_032760.1 | 1, 2 | 1118 | |||
GenBank mRNA | AF010130.1 | - | 1, 2 | 1118 | ||
AK138643.1 | - | 1, 2 | 696 | |||
BC137838.1 | - | 1, 2 | 852 | 8860857 | ||
BC144964.1 | - | 1, 2 | 855 | 9053512 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
