siRNA Details
Gene Symbol: P4hb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011032.2 | NP_035162.1 | 8 | 1238 | ||
GenBank mRNA | AK141304.1 | - | 8 | 1175 | ||
AK146542.1 | - | 8 | 1176 | |||
AK149605.1 | - | 8 | 1177 | |||
AK149965.1 | - | 8 | 1176 | |||
AK150002.1 | - | 8 | 1202 | |||
AK150422.1 | - | 8 | 1176 | |||
AK150592.1 | - | 8 | 1176 | |||
AK150755.1 | - | 8 | 1162 | |||
AK150805.1 | - | 8 | 1176 | |||
AK151351.1 | - | 8 | 1170 | |||
AK151546.1 | - | 8 | 1176 | |||
AK152141.1 | - | 8 | 1176 | |||
AK152217.1 | - | 8 | 1176 | |||
AK152791.1 | - | 8 | 1176 | |||
AK152834.1 | - | 8 | 1176 | |||
AK153365.1 | - | 8 | 1176 | |||
AK159606.1 | - | 8 | 1176 | |||
AK159729.1 | - | 8 | 1176 | |||
AK159965.1 | - | 8 | 1176 | |||
AK161537.1 | - | 8 | 1176 | |||
AK167757.1 | - | 1 | 14 | |||
AK167796.1 | - | 8 | 1176 | |||
AK168330.1 | - | 8 | 1174 | |||
AK168336.1 | - | 8 | 1158 | |||
AK168612.1 | - | 8 | 1176 | |||
AK168893.1 | - | 8 | 1176 | |||
AK169265.1 | - | 8 | 1173 | |||
AK170603.1 | - | 8 | 1173 | |||
BC008549.1 | - | 8 | 1164 | 3591404 | ||
BC018581.1 | - | 1 | 24 | |||
BC093512.1 | - | 8 | 1155 | 5124376 | ||
J05185.1 | - | 8 | 1238 | |||
X06453.1 | - | 8 | 1178 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
