siRNA Details
Gene Symbol: Reg3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011036.1 | NP_035166.1 | 3 | 191 | ||
GenBank mRNA | BC027525.1 | - | 2 | 174 | 3471932 | |
D13509.1 | - | 164 | ||||
D63359.1 | - | 3 | 191 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
