siRNA Details
Gene Symbol: Per2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011066.3 | NP_035196.2 | 21 | 3488 | ||
GenBank mRNA | AF035830.1 | - | 21 | 3435 | ||
AF036893.1 | - | 21 | 3458 | |||
AK122253.1 | - | 21 | 3453 | |||
AK159847.1 | - | 21 | 3477 | |||
AK165556.1 | - | 21 | 3491 | |||
BC055933.1 | - | 2 | 111 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
