siRNA Details
Gene Symbol: Prkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008858.3 | NP_032884.2 | 14 | 2096 | ||
GenBank mRNA | Z34524.1 | - | 14 | 1971 |

More Information
Other Species Targeted

Gene Ontology Categories:
