siRNA Details
Gene Symbol: Plcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081456.1 | NP_001074925.1 | 14, 15 | 2243 | ||
NM_148937.2 | NP_683739.2 | 15, 16 | 2490 | |||
GenBank mRNA | AK029561.1 | - | 15, 16 | 2503 | ||
AK140231.1 | - | 14, 15 | 2227 | |||
AY033991.1 | - | 15, 16 | 2322 | |||
BC066156.1 | - | 14, 15 | 2134 | 30099811 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
