siRNA Details
Gene Symbol: Plcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081456.1 | NP_001074925.1 | 6 | 797 | ||
NM_148937.2 | NP_683739.2 | 6 | 948 | |||
GenBank mRNA | AK016945.1 | - | 6 | 945 | ||
AK029561.1 | - | 6 | 943 | |||
AK039149.1 | - | 6 | 775 | |||
AK140231.1 | - | 6 | 784 | |||
AY033991.1 | - | 6 | 780 | |||
BC066156.1 | - | 6 | 688 | 30099811 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
