siRNA Details
Gene Symbol: Eif2ak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011163.4 | NP_035293.1 | 7, 8 | 824 | ||
GenBank mRNA | AK028602.1 | - | 7, 8 | 824 | ||
BC016422.1 | - | 7, 8 | 787 | 3964935 | ||
M65029.1 | - | 7, 8 | 776 | |||
M93567.1 | - | 7, 8 | 766 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
