siRNA Details
Gene Symbol: Pter | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012076.1 | - | 3 | 758 | ||
AK075604.1 | - | 3 | 757 | |||
AK135948.1 | - | 3 | 758 | |||
BC003793.1 | - | 3 | 725 | 3601675 | ||
U28016.1 | - | 3 | 734 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
