siRNA Details
Gene Symbol: Nadk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159637.1 | NP_001153109.1 | 12 | 1468 | ||
NM_138671.2 | NP_619612.2 | 12 | 1367 | |||
GenBank mRNA | AK147917.1 | - | 12 | 1445 | ||
AK164862.1 | - | 10 | 1552 | |||
AK165510.1 | - | 12 | 1461 | |||
AK170095.1 | - | 12 | 1449 | |||
AK170608.1 | - | 12 | 1463 | |||
AK170960.1 | - | 12 | 1449 | |||
AK180033.1 | - | 1 | 79 | |||
BC004012.1 | - | 12 | 1355 | 3493623 | ||
BF682935.1 | - | 2 | 197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
