siRNA Details
Gene Symbol: Ptpn20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008978.2 | NP_033004.1 | 9 | 888 | ||
GenBank mRNA | AK017003.1 | - | 9 | 951 | ||
AK029493.1 | - | 9 | 886 | |||
D64141.1 | - | 8 | 887 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
