siRNA Details
Gene Symbol: Dpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291078.1 | NP_001278007.1 | 2 | 146 | ||
NM_011262.5 | NP_035392.1 | 2 | 146 | |||
GenBank mRNA | AK004812.1 | - | 2 | 83 | ||
AK138047.1 | - | 2 | 80 | |||
AK144106.1 | - | 2 | 85 | |||
AK144848.1 | - | 2 | 81 | |||
AK144954.1 | - | 2 | 92 | |||
AK145851.1 | - | 2 | 142 | |||
AK215194.1 | - | 43 | ||||
BB839618.1 | - | 2 | 148 | |||
BC012709.1 | - | 2 | 76 | 4208205 | ||
BY294588.1 | - | 2 | 139 | |||
CX207082.1 | - | 2 | 70 | |||
U10435.1 | - | 2 | 33 | |||
U43921.1 | - | 2 | 98 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
