siRNA Details
Gene Symbol: Rlim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011276.3 | NP_035406.3 | 5 | 718 | ||
GenBank mRNA | AF069992.1 | - | 5 | 673 | ||
AK013207.1 | - | 5 | 690 | |||
AK029295.1 | - | 5 | 712 | |||
BC012960.1 | - | 3 | 451 | 4193003 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001024892, XM_003752081, XM_006257149, XM_006257141, BC097491.
Gene Ontology Categories:
