siRNA Details
Gene Symbol: Rpl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011292.2 | NP_035422.1 | 5 | 420 | ||
GenBank mRNA | AF260271.1 | - | 4 | 390 | ||
AK012331.1 | - | 5 | 390 | |||
AK012445.1 | - | 5 | 390 | |||
AK017388.1 | - | 5 | 390 | |||
AK017427.1 | - | 5 | 390 | |||
AK050296.1 | - | 2 | 1511 | |||
AK053431.1 | - | 3 | 1550 | |||
AK084278.1 | - | 5 | 391 | |||
AK088166.1 | - | 5 | 390 | |||
AK153562.1 | - | 5 | 390 | |||
AK159980.1 | - | 5 | 390 | |||
AK168326.1 | - | 5 | 388 | |||
AK169217.1 | - | 5 | 388 | |||
AK191132.1 | - | 1 | 58 | |||
BC013165.1 | - | 4 | 380 | 2655358 | ||
BC069963.1 | - | 1 | 350 | |||
BC081435.1 | - | 4 | 366 | 6824107 | ||
BC083166.1 | - | 4 | 371 | 6397429 | ||
BC083329.1 | - | 4 | 369 | 6824346 | ||
BC086937.1 | - | 5 | 387 | 6787671 | ||
BC089319.1 | - | 4 | 369 | 6823996 | ||
BC093526.1 | - | 4 | 369 | 5150493 | ||
U17331.1 | - | 3 | 326 | |||
U17332.1 | - | 3 | 326 |

More Information
Other Species Targeted

Gene Ontology Categories:
