siRNA Details
Gene Symbol: Ccl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011329.3 | NP_035459.1 | 2, 3 | 265 | ||
GenBank mRNA | AF065924.1 | - | 2, 3 | 244 | ||
AF065925.1 | - | 2, 3 | 244 | |||
AF065926.1 | - | 2, 3 | 244 | |||
AK019040.1 | - | 2, 3 | 265 | |||
BC042744.1 | - | 1876 | ||||
BC120806.1 | - | 2, 3 | 256 | 40129729 | ||
M17957.1 | - | 2, 3 | 268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
