siRNA Details
Gene Symbol: St3gal5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035228.2 | NP_001030305.1 | 2 | 212 | ||
NM_011375.3 | NP_035505.2 | 2 | 424 | |||
GenBank mRNA | AB018048.1 | - | 2 | 152 | ||
AF119416.1 | - | 2 | 202 | |||
AK012961.1 | - | 2 | 193 | |||
AK158594.1 | - | 2 | 246 | |||
AK159000.1 | - | 2 | 201 | |||
AK165726.1 | - | 2 | 214 | |||
AK217364.1 | - | 2 | 55 | |||
BB866983.1 | - | 2 | 217 | |||
BC138557.1 | - | 2 | 129 | 8861577 | ||
BC138559.1 | - | 2 | 127 | 8861579 | ||
Y15003.1 | - | 2 | 185 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
