siRNA Details
Gene Symbol: Smarcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009211.2 | NP_033237.2 | 4, 5 | 581 | ||
GenBank mRNA | AK042194.1 | - | 5, 6 | 494 | ||
AK133922.1 | - | 4, 5 | 556 | |||
AK143485.1 | - | 4, 5 | 569 | |||
AK144122.1 | - | 4, 5 | 555 | |||
AK144631.1 | - | 4, 5 | 549 | |||
AK145352.1 | - | 4, 5 | 548 | |||
BC022173.1 | - | 4, 5 | 545 | |||
BC052423.1 | - | 4, 5 | 543 | 6400646 | ||
BC053064.1 | - | 4, 5 | 537 | |||
BC056375.1 | - | 4, 5 | 544 | |||
U85614.1 | - | 4, 5 | 542 |

More Information
Other Species Targeted

Gene Ontology Categories:
