siRNA Details
Gene Symbol: Smarcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009211.2 | NP_033237.2 | 14 | 1461 | ||
GenBank mRNA | AK133922.1 | - | 14 | 1436 | ||
AK143485.1 | - | 14 | 1449 | |||
AK144122.1 | - | 14 | 1435 | |||
AK144631.1 | - | 14 | 1429 | |||
AK145352.1 | - | 14 | 1428 | |||
AK204398.1 | - | 1 | 49 | |||
BC022173.1 | - | 14 | 1425 | |||
BC052423.1 | - | 14 | 1423 | 6400646 | ||
BC053064.1 | - | 14 | 1417 | |||
BC056375.1 | - | 14 | 1424 | |||
U85614.1 | - | 14 | 1422 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
