siRNA Details
Gene Symbol: Sorl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011436.3 | NP_035566.2 | 9 | 1439 | ||
GenBank mRNA | AF031816.1 | - | 717 | |||
AK147303.1 | - | 9 | 1440 | |||
AK155045.1 | - | 9 | 1279 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
