siRNA Details
Gene Symbol: Vmn1r183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203489.1 | NP_987074.1 | 1 | 705 | ||
GenBank mRNA | BC130237.1 | - | 1 | 709 | 40142092 |
Gene Symbol: Gm10665 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167160.1 | NP_001160632.1 | 1 | 705 |
Gene Symbol: Gm10670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167161.1 | NP_001160633.1 | 1 | 705 | ||
GenBank mRNA | BC107204.1 | - | 1 | 704 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
