siRNA Details
Gene Symbol: Slc22a22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172378.2 | NP_759010.1 | 11 | 1904 | ||
GenBank mRNA | AB520670.1 | - | 10 | 1510 | ||
AK030222.1 | - | 11 | 1904 | |||
BC026439.1 | - | 10 | 1547 | 4239636 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
