siRNA Details
Gene Symbol: P3h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173379.2 | NP_775555.1 | 9, 10 | 1548 | ||
GenBank mRNA | AJ430350.1 | - | 9, 10 | 1549 | ||
AK076434.1 | - | 566 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
