siRNA Details
Gene Symbol: Themis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB509340.1 | - | 4, 5 | 1857 | ||
AK040594.1 | - | 4, 5 | 2036 | |||
AK041562.1 | - | 4, 5 | 1962 | |||
AK088136.1 | - | 4, 5 | 2038 | |||
BC125486.1 | - | 4, 5 | 1919 | 40130001 | ||
BC125488.1 | - | 4, 5 | 1919 | 40130003 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
