siRNA Details
Gene Symbol: Themis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB509340.1 | - | 4 | 1043 | ||
AK040594.1 | - | 4 | 1222 | |||
AK041386.1 | - | 4 | 1259 | |||
AK041562.1 | - | 4 | 1148 | |||
AK088136.1 | - | 4 | 1224 | |||
AK138701.1 | - | 4 | 1222 | |||
AK169558.1 | - | 5 | 1154 | |||
BC125486.1 | - | 4 | 1105 | 40130001 | ||
BC125488.1 | - | 4 | 1105 | 40130003 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
