siRNA Details
Gene Symbol: Ddx46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282055.1 | NP_001268984.1 | 16 | 2151 | ||
GenBank mRNA | AK088950.1 | - | 15 | 3741 | ||
AK129220.1 | - | 15 | 3742 | |||
AK134073.1 | - | 4 | 1397 | |||
BC026492.1 | - | 9 | 1096 | 4505095 | ||
BC092240.1 | - | 15 | 2062 | 30662573 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
