siRNA Details
Gene Symbol: Ddx46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282055.1 | NP_001268984.1 | 8 | 1113 | ||
GenBank mRNA | AK088950.1 | - | 8 | 1085 | ||
AK129220.1 | - | 8 | 1086 | |||
BC026492.1 | - | 1 | 58 | 4505095 | ||
BC092240.1 | - | 7 | 1024 | 30662573 |

More Information
Other Species Targeted

Gene Ontology Categories:
