siRNA Details
Gene Symbol: Tacr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009314.4 | NP_033340.3 | 1 | 431 | ||
GenBank mRNA | AK033443.1 | - | 1 | 114 | ||
BC106959.1 | - | 1 | 80 | 40039032 | ||
X62933.1 | - | 1 | 431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
