siRNA Details
Gene Symbol: Taf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020614.2 | NP_065639.2 | 10, 11 | 1181 | ||
GenBank mRNA | AK015111.1 | - | 10, 11 | 1148 | ||
AK131963.1 | - | 10, 11 | 1155 | |||
AK165580.1 | - | 10, 11 | 1161 | |||
AK170313.1 | - | 10, 11 | 1147 | |||
BC057167.1 | - | 10, 11 | 1168 | 4954753 | ||
BC069907.1 | - | 9, 10 | 1038 | 4951630 | ||
BC086643.1 | - | 10, 11 | 1142 | |||
BC094035.1 | - | 9, 10 | 1033 | 5029813 | ||
BC125027.1 | - | 10, 11 | 1130 | 40061781 | ||
BC125028.1 | - | 10, 11 | 1130 | 40061782 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
