siRNA Details
Gene Symbol: Tbrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130457.1 | NP_001123929.1 | 3 | 586 | ||
NM_134011.2 | NP_598772.1 | 3 | 586 | |||
GenBank mRNA | AK028966.1 | - | 3 | 564 | ||
AK129245.1 | - | 3 | 541 | |||
AK144813.1 | - | 3 | 584 | |||
AK166760.1 | - | 4 | 494 | |||
AK168088.1 | - | 3 | 584 | |||
AK188017.1 | - | 1 | 201 | |||
BC016483.1 | - | 3 | 542 | 3155271 | ||
BC031155.1 | - | 3 | 557 | 4976690 | ||
BC031910.1 | - | 3 | 556 | 5362943 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
