siRNA Details
Gene Symbol: Ago3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153402.2 | NP_700451.2 | 9 | 1376 | ||
GenBank mRNA | AB081473.1 | - | 9 | 1228 | ||
AK035042.1 | - | 9 | 1342 | |||
AK083652.1 | - | 9 | 1343 | |||
AK154436.1 | - | 9 | 1377 | |||
AK171118.1 | - | 9 | 1377 | |||
BC137964.1 | - | 9 | 1223 | 8860983 | ||
BC145061.1 | - | 9 | 1223 | |||
BC145063.1 | - | 8 | 1086 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001271193, XM_008764023, XM_008764024.
Gene Ontology Categories:
