siRNA Details
Gene Symbol: Tmem51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145402.3 | NP_663377.1 | 3 | 1630 | ||
GenBank mRNA | AK077758.1 | - | 3 | 1630 | ||
AK164026.1 | - | 3 | 1572 | |||
AK189441.1 | - | 1 | 38 | |||
AK195248.1 | - | 1 | 38 | |||
BC003277.1 | - | 3 | 1494 | 3582174 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
