siRNA Details
Gene Symbol: Arrdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162485.1 | NP_001155957.1 | 6 | 918 | ||
NM_178408.3 | NP_848495.2 | 6 | 915 | |||
GenBank mRNA | AK088008.1 | - | 6 | 745 | ||
AK170018.1 | - | 6 | 792 | |||
BC004091.1 | - | 6 | 813 | |||
BC018501.1 | - | 6 | 797 | 3662534 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
