siRNA Details
Gene Symbol: Ugp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290634.1 | NP_001277563.1 | 8 | 1327 | ||
NM_139297.6 | NP_647458.1 | 8 | 1517 | |||
GenBank mRNA | AF424698.1 | - | 8 | 1365 | ||
AK033445.1 | - | 8 | 1364 | |||
AK153885.1 | - | 7 | 2460 | |||
BC023810.1 | - | 8 | 1368 | 5353993 | ||
BC025585.1 | - | 8 | 1272 | 5324928 | ||
BC026626.1 | - | 7 | 1054 | |||
BC061208.1 | - | 8 | 1404 | 30250244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
