siRNA Details
Gene Symbol: Ugp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290634.1 | NP_001277563.1 | 10 | 1711 | ||
NM_139297.6 | NP_647458.1 | 10 | 1901 | |||
GenBank mRNA | AF424698.1 | - | 10 | 1749 | ||
AK033445.1 | - | 10 | 1748 | |||
AK153885.1 | - | 9 | 2844 | |||
AK177417.1 | - | 1 | 74 | |||
AK205646.1 | - | 3 | 383 | |||
AK211484.1 | - | 3 | 379 | |||
AK215699.1 | - | 3 | 361 | |||
AK216282.1 | - | 3 | 351 | |||
AK217204.1 | - | 3 | 382 | |||
AK220026.1 | - | 3 | 376 | |||
BC023779.1 | - | 3 | 350 | |||
BC023810.1 | - | 10 | 1752 | 5353993 | ||
BC025585.1 | - | 10 | 1656 | 5324928 | ||
BC026626.1 | - | 9 | 1438 | |||
BC061208.1 | - | 10 | 1788 | 30250244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
