siRNA Details
Gene Symbol: Kdm6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017426.1 | NP_001017426.1 | 12, 13 | 3973 | ||
GenBank mRNA | AK129121.1 | - | 2, 3 | 762 | ||
AK134236.1 | - | 2, 3 | 1094 | |||
AK136019.1 | - | 2, 3 | 154 | |||
BC075632.1 | - | 13, 14 | 4149 | 30362899 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
