siRNA Details
Gene Symbol: Camta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190376.1 | NP_001177305.1 | 8 | 908 | ||
NM_001190378.1 | NP_001177307.1 | 8 | 908 | |||
NM_001190379.1 | NP_001177308.1 | 7 | 836 | |||
NM_178116.4 | NP_835217.1 | 8 | 923 | |||
GenBank mRNA | AK039670.1 | - | 8 | 735 | ||
AK122400.1 | - | 6 | 564 | |||
AK150186.1 | - | 8 | 760 | |||
AK169118.1 | - | 7 | 675 | |||
BC023962.1 | - | 3 | 722 | |||
BC049133.1 | - | 8 | 752 | 6827800 | ||
BC056395.1 | - | 8 | 737 | 6852349 | ||
BC059065.1 | - | 7 | 703 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
