siRNA Details
Gene Symbol: Camta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190376.1 | NP_001177305.1 | 21 | 3633 | ||
NM_001190378.1 | NP_001177307.1 | 20 | 3612 | |||
NM_001190379.1 | NP_001177308.1 | 19 | 3540 | |||
NM_178116.4 | NP_835217.1 | 21 | 3648 | |||
GenBank mRNA | AK122400.1 | - | 19 | 3289 | ||
AK150186.1 | - | 20 | 3466 | |||
AK169118.1 | - | 19 | 3379 | |||
BC023962.1 | - | 15 | 3426 | |||
BC027039.1 | - | 10 | 1357 | |||
BC038079.1 | - | 3 | 239 | |||
BC049133.1 | - | 21 | 3477 | 6827800 | ||
BC056395.1 | - | 21 | 3462 | 6852349 | ||
BC059065.1 | - | 19 | 3407 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001105801, XM_006246665.
Gene Ontology Categories:
