siRNA Details
Gene Symbol: Nags | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145829.2 | NP_665828.1 | 6, 7 | 1508 | ||
GenBank mRNA | AF462069.1 | - | 6, 7 | 1464 | ||
AJ489814.1 | - | 6, 7 | 1434 | |||
AK075765.1 | - | 5, 6 | 1154 | |||
BC036161.1 | - | 3, 4 | 2159 | |||
BC057990.1 | - | 6, 7 | 1472 | 5100021 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
