siRNA Details
Gene Symbol: Nags | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145829.2 | NP_665828.1 | 4 | 1129 | ||
GenBank mRNA | AF462069.1 | - | 4 | 1085 | ||
AJ489814.1 | - | 4 | 1055 | |||
AK075765.1 | - | 3 | 775 | |||
BC036161.1 | - | 2 | 1515 | |||
BC057990.1 | - | 4 | 1093 | 5100021 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
