siRNA Details
Gene Symbol: Tff2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009363.3 | NP_033389.2 | 3, 4 | 414 | ||
GenBank mRNA | AK003093.1 | - | 3, 4 | 415 | ||
BC050086.1 | - | 3, 4 | 375 | |||
BC087889.1 | - | 3, 4 | 392 | 30316277 | ||
X51697.1 | - | 3, 4 | 379 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
